Ion Ampliseq™ Cancer Hotspot Panel v2 Chef-Ready Kit
Ion Torrent™

Ion Ampliseq™ Cancer Hotspot Panel v2 Chef-Ready Kit

The Ion Ampliseq™ Cancer Hotspot Panel v2 Chef-Ready Kit combines a single pool of primers with the reagents necessary toRead more
Have Questions?
Catalog NumberQuantity
A3291432 Reactions
Catalog number A32914
Price (USD)
10,140.00
Each
Quantity:
32 Reactions
Price (USD)
10,140.00
Each

The Ion Ampliseq™ Cancer Hotspot Panel v2 Chef-Ready Kit combines a single pool of primers with the reagents necessary to prepare libraries of frequently mutated regions in human cancer genes on the Ion Chef™ System. This automated workflow utilizes the same chemistry as the manual library construction workflow. It provides equivalent or better performance with exceptional reproducibility, on-target reads, and sample balance. Automated production of libraries using the Ion Chef System reduces hands-on time to less than 15 minutes.

The Ion Ampliseq Cancer Hostpot Panel v2 Chef Ready Kit consists of the following components:

  • Ion Ampliseq™ Cancer Hotspot Research Panel Primers
  • Ion Ampliseq™ Kit for Chef DL8

The simple sample-to-data workflow can be executed in less than two days and offers the following features and benefits:

  • Production of high-quality DNA libraries using the Ion Chef System
  • Proven performance of Ion Ampliseq™ technology, FFPE compatibility, uniform coverage for efficient sequencing, accurate variant calling, and improved variant detection via Torrent Suite™ Software
  • Ability to scale and multiplex on Ion™ chips based on desired throughput needs, driving cost savings and efficiency
  • Simplified workflow and reduced hands-on time when used with the Ion Ampliseq™ Direct FFPE DNA Kit
  • Low input amount of only 1 ng per sample

Additional mutational content for advancement of cancer research studies

The Ion AmpliSeq Cancer Hotspot Panel v2 is designed to amplify 207 amplicons covering approximately 2,800 COSMIC mutations from 50 oncogenes and tumor suppressor genes. The panel should be used with the Ion AmpliSeq™ Library Kit 2.0.

Minimal hands-on time

Leveraging the automation capabilities of the Ion Chef System, the included Ion AmpliSeq Kit for Chef Kit DL8 enables the reproducible preparation of Ion AmpliSeq amplicon libraries, with less than 15 minutes of hands-on time and only 3 liquid transfer steps. Compatible with 1- and 2-pool Ion AmpliSeq panel designs, the kit automates the preparation of libraries from 8 samples in a single run, delivering a single tube of pooled libraries ready for downstream templating and sequencing. The kit provides sufficient material for the preparation of 32 libraries.

Simplicity and ease-of-use of Ion solutions

Utilizing plug and play, pre-packaged, single-use cartridges and PCR plates for a fully automated workflow, users of all experience levels will find library preparation straightforward with minimal effort. Supplied with barcodes conveniently dried-down in 96-well plates, multiplexing samples is easy and requires no additional pipetting steps.

Specifications
For Use With (Application)Oncology Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of SamplesUp to 32 Samples
Nucleic Acid TypeDNA
Product LineIon AmpliSeq
Product TypePanel
Quantity32 Reactions
Sample TypeFFPE Tissue, Whole Blood, Cell-Free DNA
SoftwareTorrent Suite Software
ThroughputUp to 4 Samples per Ion 510 Chip, Up to 8 Samples per Ion 520 Chip, Up to 26 Samples per Ion 530 Chip
Workflow StepPanel and Library
Format96-well Plate
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.