With the Oncomine Immune Response Research Assay, how many samples can I multiplex per run?
Multiplexing can be done with 1-4 samples on the PGM 318 or S5/S5XL 520 chips, or with 2-8 samples on the S5/S5XL 520 chip. This enables flexibility for small-to-large retrospective profiling studies.
Which RNA isolation method do you recommend for use with the Oncomine Immune Response Research Assay?
We recommend using the RecoverAll Total Nucleic Acid Isolation Kit for FFPE (Cat. No. AM1975).
Can I automate the Oncomine Immune Response Research Assay workflow?
Yes, automated library and template preparation are enabled with the Ion Chef System.
Where can I find the User Guide for the Oncomine Immune Response Research Assay?
The User Guide can be found here (https://tools.thermofisher.com/content/sfs/manuals/MAN0015867_OncomineImmuneRespnseResrchAssay_UG.pdf).
What functional annotation groups are covered by the Oncomine Immune Response Research Assay?
The Oncomine Immune Response Research Assay content includes 395 genes across 36 functional annotation groups. These groups comprise genes associated with lymphocyte regulation, cytokine signaling, lymphocyte markers, checkpoint pathways, and tumor characterization.
How many genes are covered in the Oncomine Immune Response Research Assay?
The Oncomine Immune Response Research Assay covers 395 genes.
What is the minimum input amount of RNA for the Oncomine Immune Response Research Assay?
For the Oncomine Immune Response Research Assay, the minimum input amount of RNA is 10 ng.
How do I get the Oncomine Immune Response Research Plugin?
The Oncomine Immune Response Research Plugin is available in the Torrent Suite Software (TSS) as an update to TSS v5.2.1.