Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs
Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs
View additional product information for Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs (A32931TR, A32931)
1 product FAQs found
What outputs does the Oncomine Immune Response Research Plugin produce?
The plugin produces the following files:
-Summary report for all barcodes or samples processed in the same run
-Absolute read counts
-RPM data
-Expression data normalized by mean expression of housekeeping genes (scaled log2)
-CHP files derived from RPM
-CHP files derived from data normalized with housekeeping genes
-Background expression data from negative controls, genomic DNA, & H20