Can I download graphs/visualizations from the Oncomine Immune Response Research Plugin or Transcriptome Analysis Console (TAC) software?
The visualizations/graphs from the Torrent Suite Software plugin can be saved as images by right-clicking on the image and saving the image to your selected folder. The TAC also provides similar export options for the graphs to be downloaded to your computer.