Alexa Fluor™ 568 Carboxylic Acid, tris(triethylammonium) salt, 5 mg - FAQs
Alexa Fluor™ 568 Carboxylic Acid, tris(triethylammonium) salt, 5 mg - FAQs
View additional product information for Alexa Fluor™ Carboxylic Acid, tris(triethylammonium) salts - FAQs (A33079, A33078, A33080, A33082, A33081, A33084, A33083, A33085, A33077, A33076)
5 product FAQs found
What dye can I use that is non-reactive and can show an injection site?
The non-reactive Alexa Fluor and Alexa Fluor hydrazide derivatives may be used for injection site visualization. Other options include the fluorescent polystyrene microspheres, FluoSpheres, and dye-conjugated dextrans. The hydrazide derivatives and 'fixable' dextrans are retained by cross-linking using an aldehyde-based fixative.
Which optical filters are compatible with my Alexa Fluor free acid dye?
Please consult the Fluorescence SpectraViewer to find the optimal optical filters to use with your Alexa Fluor free acid dye (Cat. Nos. A33076, A33077, A33078, A33079, A33080, A33081, A33082, A33083, A33084, A33085).
For more information on how to use the Fluorescence SpectraViewer tool, please refer to the Guide for Using the Fluorescence SpectraViewer.
How should I reconstitute and store my Alexa Fluor free acid dye?
Alexa Fluor free acid compounds (Cat. Nos. A33076, A33077, A33078, A33079, A33080, A33081, A33082, A33083, A33084, A33085) should be stored at -20 degrees C and protected from light until it is reconstituted.
To reconstitute, dissolve the Alexa Fluor free acid compound in aqueous buffer (e.g., PBS), or high-quality, anhydrous dimethylsulfoxide (DMSO) or dimethylformamide (DMF) at a final concentration of 1–10 mg/mL. Once reconstituted, this solution is stable for 1 year, when stored at 2-8 degrees C and protected from light.
How long after injection should I start imaging my mouse?
The imaging time course varies with the nature of the injected agent. Vascular tracers are visible in the blood vessels immediately after injection and may be imaged for several hours. Conjugated whole IgG antibodies reach their targets within a few hours of injection and may be imaged for several days.
What happens to unconjugated dye in the mouse after small animal in vivo imaging?
The dye will be eliminated via the bladder. The bladder signal is detectable within ~3 minutes of IV injection of the dye and clearance with ~30 minutes.