Precision ID GlobalFiler™ NGS STR Panel v2 - FAQs
Precision ID GlobalFiler™ NGS STR Panel v2 - FAQs
View additional product information for Precision ID GlobalFiler™ NGS STR Panel v2 - FAQs (A33114)
4 product FAQs found
What are the expected library lengths with the Precision ID GlobalFiler STR NGS v2?
The expected library length ranges from ˜129-303 bp.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many reactions can be performed with the Precision ID GlobalFiler STR NGS v2 panel?
The Precision ID GlobalFiler STR NGS v2 panel provides enough reagents for 96 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Which Ion systems can the Precision ID GlobalFiler STR NGS panel v2 be used with?
The Precision ID GlobalFiler STR NGS panel v2 is only compatible with the Ion S5 System. It is not designed for use with the Ion OneTouch2 System or Ion PGM System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Precision ID GlobalFiler STR NGS panel v2, how many samples can be multiplexed?
Up to 32 libraries can be multiplexed on an Ion 530 chip (Cat. No. A27764) with the Precision ID GlobalFiler STR NGS panel v2.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.