MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit - FAQs
MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit - FAQs
View additional product information for MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit - FAQs (A33246)
7 product FAQs found
How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) stable for?
The product is guaranteed stable for one year when stored properly at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How many sequencing samples is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) sufficient for?
The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit is sufficient for 96 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What are the catalog numbers for the MicroSEQ ID Ultra Sequencing Clean-up Kits?
Here the kits with the catalog numbers:
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is included in the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)?
The kit consists of twelve 8-well strips and a 96-well receiver plate to accommodate 96 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips good for use once opened?
Strips should be used within 2 weeks of opening. We recommend returning unused strips from the 96-well plate holder to the foil package, sealing, and storing at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the minimum reaction volume for use of the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)?
The minimum reaction volume is 5 µL. The reaction volume should be brought up to at least 5 µL with distilled water before adding to the 8-well strips.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the storage condition for the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)?
The kit is stable when stored for 12 months at 4 degrees C or for 4 weeks at ambient temperature (20-24 degrees C).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.