What is the master mix used in the VetMAX PED/TGEV/SDCoV Kit?
The master mix used in this kit is the TaqMan Fast Virus 1-Step Master Mix.
What are the probes included in the VetMAX PEDV/TGEV/SDCoV Kit?
The TaqMan assay for PEDV RNA is labeled with LIZ, the TaqMan assay for TGEV is labeled with FAM, the TaqMan assay for SDCoV is labeled with VIC, and the TaqMan assay for the Xeno RNA is labeled with NED. All four TaqMan assays are combined in the VetMAX PEDV/TGEV/SDCoV Primer Probe Mix.
What is the master mix used in the VetMAX PEDV/TGEV/SDCoV Kit?
The master mix used in the VetMAX PEDV/TGEV/SDCoV Kit is the TaqMan Fast Virus 1-Step Master Mix.