Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate - FAQs
Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate - FAQs
View additional product information for Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate - FAQs (A33586)
3 product FAQs found
What is the volume of each barcode provided, per well, in the Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate?
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate contains 20 µL/well of each barcode.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many reactions are available per barcode with the Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate?
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate provides 10 reactions per barcode.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many reactions can be performed with the Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate (Cat. No. A33586)?
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate provides enough reagents for 960 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.