SeqStudio™ Cartridge - FAQs
SeqStudio™ Cartridge - FAQs
View additional product information for SeqStudio™ Cartridge - FAQs (A33671)
5 product FAQs found
How do I recycle my SeqStudio cartridge if my country is not supported by the recycling program?
We would recommend disposing of your SeqStudio cartridge according to your country's regulations on proper disposal of laboratory equipment and reagents.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
How do I know if the SeqStudio cartridge recycling program is supported in my country?
You can view the details and availability of the SeqStudio Cartridge recycling program on this web page (https://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/sanger-sequencing-technology-accessories/seqstudio-genetic-analyzer/seqstudio-cartridge-recycling-program.html) or speak to your local representative for more information.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Where does the recycling of my used SeqStudio cartridge take place?
We have partnered with E-End Solutions in North America and SIMMs Recycling Solutions in Europe for recycling of used SeqStudio cartridges. More details about the program can be found in the SeqStudio Cartridge Green Fact Sheet (https://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/seqstudio-cartridge-green-fact-sheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What should I do if I lost or misplaced my SeqStudio cartridge recycling return envelope?
If the original SeqStudio cartridge recycling return envelope is not available, you can place the return label on another polyethylene envelope of similar dimensions. The envelope dimensions are 12" x 15 1⁄2".
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What is the cost to recycle my SeqStudio cartridge?
There is no cost to recycle your SeqStudio cartridge when you use the pre-paid label provided by Thermo Fisher Scientific.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.