MagMAX™ Plant RNA Isolation Kit, 384 Preps - FAQs
MagMAX™ Plant RNA Isolation Kit, 384 Preps - FAQs
View additional product information for MagMAX™ Plant RNA Isolation Kit - FAQs (A33784, A33899, A47157)
3 product FAQs found
How should I prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899, A47157) DNase I solution?
To prepare the MagMAX Plant RNA Kit (Cat. Nos. A33784, A33899) DNase I solution, add 440 µL of DNase I Reconstitution Buffer to each vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.
To prepare the MagMAX Plant RNA Kit (Cat. No. A47157) DNase I solution, add 10.5 mL of DNase I Reconstitution Buffer to the vial of DNase I (lyophilized) and incubate at room temperature for 5 minutes.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
What kind of Polyvinylpyrrolidone (PVP) do you recommend using with MagMAX Plant DNA and RNA isolation kits since there are different kinds available?
For the MagMAX Plant DNA Isolation Kit, we recommend using PVP M.W. 40,000, Cat. No. J62417.A1 (https://www.thermofisher.com/order/catalog/product/J62417.A1).
For the MagMAX Plant RNA Isolation Kit, we recommend using Plant RNA Isolation Aid, Cat. No. AM9690 (https://www.thermofisher.com/order/catalog/product/AM9690).
Can I use the MagMAX Plant RNA Isolation Kit (Cat. No. A33784) to isolate small RNAs?
The MagMAX Plant RNA Isolation Kit has not been optimized for the isolation of small RNAs.