Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
AgriSeq™ Normalization Reagents Kit
Applied Biosystems™

AgriSeq™ Normalization Reagents Kit

The Applied Biosystems AgriSeq Normalization Reagents Kit is specifically designed for high-throughput normalization of amplicon libraries for targeted genotyping-by-sequencing (GBS) applications in agrigenomics.
Have Questions?
Catalog NumberQuantity
A341409600 Reactions
Catalog number A34140
Price (USD)
-
Quantity:
9600 Reactions
The Applied Biosystems AgriSeq Normalization Reagents Kit is specifically designed for high-throughput normalization of amplicon libraries for targeted genotyping-by-sequencing (GBS) applications in agrigenomics.

Key product features include:

  • Exceptional uniformity of multiplexed target amplification facilitate robust, reproducible results
  • Simplified workflow enables high-throughput sample processing
  • Convenient configuration contains library normalization reagents and volumes sufficient for automated platforms
  • Compatible with AgriSeq HTS and AgriSeq ULTRA workflows

Conveniently packaged normalization reagents and volumes for automation

The AgriSeq Normalization Reagents Kit is offered in a large pack size (9600 reactions) to accommodate high-throughput agrigenomics projects. The kit includes all reagents required for library normalization to help achieve consistent results between samples, and can accommodate automated workflows. AgriSeq targeted GBS is an economical way to generate high-throughput genotyping data to help advance your breeding programs.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion Torrent
No. of Reactions9600
Quantity9600 Reactions
FormatKit
Sequencing TypeNext Generation Sequencing
Unit Size9600 reactions