Product Image
Ion Torrent™
The Ion Chef™ Calibration Supplies Kit
The Ion Chef Calibration Supplies Kit is used to perform the XYθ and Z calibration routines on the Ion Chef Instrument.
Have Questions?
Catalog NumberQuantity
A343011 kit
Catalog number A34301
Price (USD)
540.00
Each
Quantity:
1 kit
Price (USD)
540.00
Each
The Ion Chef Calibration Supplies Kit is used to perform the XYθ and Z calibration routines on the Ion Chef Instrument. These routines find the precise location of the tip mandrel relative to each consumable cartridge and a home position for the three centrifuges.
Features include:
  • The ability to recalibrate your Ion Chef Instrument without having to open a service order
  • No more waiting for service engineers to perform a calibration
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
Product LineIon Chef
Product TypeCalibration Supplies Kit
Quantity1 kit
Shipping ConditionRoom Temperature
FormatKit
Unit SizeEach
Contents & Storage
• 10 Ion Chef PCR plates
• 96 Black tips, 1 mL
• 12 Ion Chef recovery tubes
• 10 Pierce tools
• 2 Ion Chef tip cartridges
• 2 Ion Chef tubs