TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast - FAQs
TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast - FAQs
View additional product information for TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast - FAQs (A34643)
4 product FAQs found
Can the TaqMan Advanced miRNA Human Endogenous Controls plate (Cat. No. A34643) which has pre-spotted TaqMan Advanced Assays be used for mouse samples?
While a lot of targets between human, mouse and rat are the same we had to design specific assays for mouse and rat because cross-reactivity tests are done within a species to exclude off-targets. Assays on the human endogenous control plate will detect the corresponding mouse and rat targets, however it is possible that some human assays might detect off-targets in mouse.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?
The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.
Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?
We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.
Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch
How do I obtain TaqMan documentation files for my TaqMan array plates?
Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number