POROS™ Benzyl Ultra RoboColumn™ Pre-packed columns
Thermo Scientific™

POROS™ Benzyl Ultra RoboColumn™ Pre-packed columns

This Thermo Scientific™ RoboColumn™ product is pre-packed with POROS Benzyl Ultra Hydrophobic Interaction Chromatography (HIC) Resin that is well suited for aggregate removal from monoclonal antibody monomer in flow-through mode at low salt concentrations in large scale downstream bioprocessing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A348161 Strip, 8 Columns
A348151 Strip, 8 Columns
Catalog number A34816
Price (USD)
363.00
Each
Quantity:
1 Strip, 8 Columns
Price (USD)
363.00
Each

This RoboColumn™ product is pre-packed with POROS Benzyl Ultra Hydrophobic Interaction Chromatography (HIC) Resin (Cat. No. A32564) that is well suited for aggregate removal from monoclonal antibody monomer in flow-through mode at low salt concentrations in large scale downstream bioprocessing.

RoboColumn™ products are small chromatography columns that are provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.Features of POROS Benzyl Ultra HIC Resin include:

  • Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50-micron particle size. Higher flow rates help improve productivity.
  • Unique selectivity due to the novel hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
  • High capacity, reducing the process volume significantly for subsequent steps
  • A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process

These features of POROS Benzyl Ultra HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography.

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications
Bed Volume600 μL
Column Capacity600 μL
Column FormatPre-packed Robo Column
For Use With (Equipment)Robotic Fluid Handlers
MatrixCross-linked poly(styrene-divinylbenzene) POROS beads
Particle Size50 μm
Quantity1 Strip, 8 Columns
Shipping ConditionAmbient
Stationary PhaseHigh-Density Benzyl/Aromatic
Diameter (Metric) Inner0.5 cm
Length (Metric)3 cm
Pore SizeNot Applicable
Product LinePOROS
TypeHIC Column
Unit SizeEach
Contents & Storage
Store at 2°C to 30°C. Do not freeze.