ReproSeq™ PGS Kit with Ion 510™ Chips (16 samples/run) - FAQs
ReproSeq™ PGS Kit with Ion 510™ Chips (16 samples/run) - FAQs
View additional product information for ReproSeq™ PGS Kit with Ion 510™ Chips (16 samples/run) - FAQs (A34899)
3 product FAQs found
How many total samples can I process with the Ion ReproSeq PGS Kit with Ion 510 Chips?
64 samples in total (4 runs with 16 samples per run) can be processed with the Ion ReproSeq PGS Kit with Ion 510 Chips.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many samples per run can I process with the Ion ReproSeq PGS Kit with Ion 510 Chips?
16 samples per run can be processed with the Ion ReproSeq PGS Kit with Ion 510 Chips.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What does the Ion ReproSeq PGS Kit with Ion 510 Chips (16 samples/run) include, and what are the storage conditions for each component?
*Ion SingleSeq Kit, 3 boxes, -10 to -30 degrees C
*Ion S5 ExT Chef Supplies, 4 boxes, 15-30 degrees C
*Ion S5 Chef Solutions, 4 cartridges, 15-30 degrees C
*Ion S5 ExT Chef PGS Reagents, 4 cartridges, -10 to -30 degrees C
*Ion S5 ExT Sequencing Solutions, 1 box, 15-30 degrees C
*Ion S5 ExT Sequencing Reagents, 4 cartridges, -10 to -30 degrees C
*Ion 510 Chip Kit, 4 chips, 15-30 degrees C
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.