C57BL/6 Mouse Embryonic Fibroblasts, irradiated
Gibco™
C57BL/6 Mouse Embryonic Fibroblasts, irradiated
Irradiated Gibco C57BL/6 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that support the culture of healthy undifferentiated embryonic stem cellsRead more
Have Questions?
Catalog NumberNo. of Cells
A349614 x 106 cells
Catalog number A34961
Price (TWD)
2,373.00
Online offer
Ends: 30-Jun-2026
3,390.00
Save 1,017.00 (30%)
Each
No. of Cells:
4 x 106 cells
Price (TWD)
2,373.00
Online offer
Ends: 30-Jun-2026
3,390.00
Save 1,017.00 (30%)
Each
Irradiated Gibco C57BL/6 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that support the culture of healthy undifferentiated embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs), especially mouse ESCs/iPSCs. Manufactured by MTI-GlobalStem and equivalent to MTI-GlobalStem Cat. Nos. GSC-6002G and GSC-6202G, these MEF cells are isolated from inbred C57BL/6 mice, mitotically arrested by irradiation, and tested to help ensure performance and minimize the risk of contamination.
• Help save precious time with pre-inactivated, ready-to-use MEFs
• Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
• Provide optimum support for mouse ESCs and iPSCs
Help save time with ready-to-use MEFs
Irradiated C57BL/6 MEFs help save time and trouble dealing with an animal facility, performing tedious, time-consuming dissections, and expanding and testing cells day after day, week after week. Focus on your research with these pre-inactivated, ready-to-use feeders.
Culture with meticulously tested feeder cells
Each lot of irradiated C57BL/6 MEFs is tested for bacteria, fungi, mycoplasma, and mouse pathogens to minimize the risk of contamination. Each lot is also tested for post-thaw viability (typically greater than 95%) and ability to support mouse and human ESCs to help ensure robust and consistent performance. Culture your ESCs and iPSCs with confidence, knowing that you can rely on these high quality feeder cells.
Provide optimum support for mouse ESCs and iPSCs
While C57BL/6 MEFs can also support human ESCs and iPSCs, these feeder cells work particularly well for culturing mouse ESCs and iPSCs, showing better results compared to CF1 and ICR strain MEFs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor SourcePooled
No. of Cells4 x 106 cells
Quantity4 x 106 cells
Shipping ConditionShipped on dry ice
AgeEmbryonic
Cell TypeFeeder Cells (Mouse Embryonic Fibroblast)
FormCryopreserved
SpeciesMouse
Unit SizeEach
Contents & Storage
Cryopreserved mouse embryonic fibroblasts
Store in liquid nitrogen.
Frequently asked questions (FAQs)
Is it better to use irradiated mouse embryonic fibroblasts (MEFs) or Mitomycin C-treated MEFs to grow embryonic stem cells?
Either method will work in arresting cell division. However, the irradiation process will ensure that cell division will cease regardless of cell aggregation. Cell clumping can potentially not inactivate all cells when using mitomycin C, as cells within clumps may not be exposed to the mitomycin C. Irradiated cells are preferred by those who have concerns about chemical treatment. Mitomycin C-treated cells are preferred by those who have concerns about DNA damage from irradiation.
What is the recommended seeding density for mouse embryonic fibroblasts? Why is the seeding density critical?
We recommend seeding these cells at densities ranging from 2 x 10E4 to 5.5 x 10E4 cells/cm2. A good starting point is 3 x 10E4 cells/cm2. If the feeder cells are too sparse, they may not maintain the pluripotent cells without differentiation, and the pluripotent cells may not attach well. If the feeder cells are too dense, the feeder layer may detach from the plate, and the culture will be lost.
When should mouse embryonic fibroblasts be plated and how long can these cells be used?
These cells should be plated 24 hours prior to plating the ESCs or iPSCs and should be used for only 7-10 days.
Does the culture vessel need to be coated prior to seeding mouse embryonic fibroblasts?
Yes, the culture vessel needs to be coated with Attachment Factor protein (Cat. No. S006100) at 37 degrees C for 30 mins or at room temperature for 2 hours. The coated vessels can be used immediately or stored at room temperature for up to 24 hours.