Use with Annexin Binding Buffer for flow cytometry
Annexin V Conjugates for Apoptosis Detection
Invitrogen™

Annexin V Conjugates for Apoptosis Detection

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/EmissionFlow Cytometer Laser LinesConjugate
A35110650/660633-637APC (Allophycocyanin)
A13201495/519488Alexa Fluor 488
A13199494/518488FITC
A13202578/603532, 561Alexa Fluor 568
A13203590/617532Alexa Fluor 594
A13204Biotin-X
A23202346/442UVAlexa Fluor 350
A23204650/665633-637Alexa Fluor 647
A35108555/565532, 561Alexa Fluor 555
A35109679/702633-637Alexa Fluor 680
A35111565/578488, 532, 561PE
A35122410/455405Pacific Blue
Catalog number A35110
Price (USD)
582.00
Each
Excitation/Emission:
650/660
Flow Cytometer Laser Lines:
633-637
Conjugate:
APC (Allophycocyanin)
Price (USD)
582.00
Each
Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.
Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
DescriptionAnnexin V, APC conjugate (APC Annexin V) (replaces Annexin product- AnnexinV05)
Excitation/Emission650/660
Flow Cytometer Laser Lines633-637
For Use With (Equipment)Flow Cytometer
No. of Reactions50
Product TypeAnnexin V conjugate
Quantity250 μL
Shipping ConditionWet Ice
ConjugateAPC (Allophycocyanin)
Unit SizeEach
Contents & Storage
Contains 1 vial of annexin V, APC conjugate. Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

When should I use Annexin V instead of an Invitrogen CellEvent Caspase Detection Reagent?

Choose Annexin V to detect phosphatidylserine exposure at the plasma membrane, and choose Invitrogen CellEvent Caspase Detection Reagents to detect activated caspases and nuclear fluorescence. The methods measure different apoptotic events and should not be treated as interchangeable.

I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I fix my cells after annexin V staining?

Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.

Citations & References (15)

Citations & References
Abstract
Simultaneous detection of apoptosis and mitochondrial superoxide production in live cells by flow cytometry and confocal microscopy.
Authors:Mukhopadhyay P, Rajesh M, Haskó G, Hawkins BJ, Madesh M, Pacher P
Journal:Nat Protoc
PubMed ID:17853886
'Annexin V and Sytox Green are widely used markers to evaluate apoptosis in various cell types using flow cytometry and fluorescent microscopy. Recently, a novel fluoroprobe MitoSOX Red was introduced for selective detection of superoxide in the mitochondria of live cells and was validated for confocal microscopy and flow cytometry. ... More
Cathepsin D and H2O2 stimulate degradation of thioredoxin-1: implication for endothelial cell apoptosis.
Authors:Haendeler J, Popp R, Goy C, Tischler V, Zeiher AM, Dimmeler S
Journal:J Biol Chem
PubMed ID:16263712
'Cathepsin D (CatD) is a lysosomal aspartic proteinase and plays an important role in the degradation of proteins and in apoptotic processes induced by oxidative stress, cytokines, and aging. All of these stimuli are potent inducers of endothelial cell apoptosis. Therefore, we investigated the role of CatD in endothelial cell ... More
Role of superoxide, nitric oxide, and peroxynitrite in doxorubicin-induced cell death in vivo and in vitro.
Authors:Mukhopadhyay P, Rajesh M, Bátkai S, Kashiwaya Y, Haskó G, Liaudet L, Szabó C, Pacher P,
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:19286953
'Doxorubicin (DOX) is a potent available antitumor agent; however, its clinical use is limited because of its cardiotoxicity. Cell death is a key component in DOX-induced cardiotoxicity, but its mechanisms are elusive. Here, we explore the role of superoxide, nitric oxide (NO), and peroxynitrite in DOX-induced cell death using both ... More
FTY720 increases CD74 expression and sensitizes mantle cell lymphoma cells to milatuzumab-mediated cell death.
Authors:Alinari L, Mahoney E, Patton J, Zhang X, Huynh L, Earl CT, Mani R, Mao Y, Yu B, Quinion C, Towns WH, Chen CS, Goldenberg DM, Blum KA, Byrd JC, Muthusamy N, Praetorius-Ibba M, Baiocchi RA,
Journal:Blood
PubMed ID:22042694
'Mantle cell lymphoma (MCL) is an aggressive B-cell malignancy with a short median survival despite multimodal therapy. FTY720, an immunosuppressive drug approved for the treatment of multiple sclerosis, promotes MCL cell death concurrent with down-modulation of phospho-Akt and cyclin D1 and subsequent cell-cycle arrest. However, the mechanism of FTY720-mediated MCL ... More
Flow cytometry-based apoptosis detection.
Authors:Wlodkowic D, Skommer J, Darzynkiewicz Z,
Journal:Methods Mol Biol
PubMed ID:19609746
'An apoptosing cell demonstrates multitude of characteristic morphological and biochemical features, which vary depending on the stimuli and the cell type. The gross majority of classical apoptotic hallmarks can be rapidly examined by flow and image cytometry. Cytometry thus became a technology of choice in diverse studies of cellular demise. ... More