Calf Serum with Iron (US Origin) - FAQs
Calf Serum with Iron (US Origin) - FAQs
View additional product information for Calf Serum with Iron (US Origin) - FAQs (A3520502)
2 product FAQs found
What is the advantage of using Calf Serum with Iron (US Origin)?
The addition of iron in calf serum can result in higher performance of serum in cell culture applications. Iron-supplemented calf serum can be a viable alternative to fetal bovine serum in the growth of certain cells. Calf Serum with Iron (US Origin) is collected from calves 1-6 months of age, which naturally have higher levels of transferrin to bind iron. The iron content in Calf Serum with Iron (US Origin) is 500-700 µg/dL.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of Calf Serum with Iron (US Origin)?
The shelf life is 48 months from the date of manufacture.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.