Attune™ NxT Automation Bundle, stacks/hotel
Invitrogen™
Attune™ NxT Automation Bundle, stacks/hotel
Automate your flow cytometry workstation with the Attune NxT and Orbitor RS2 Automation Bundle. The Orbitor RS2 Microplate Mover isRead more
Have Questions?
Catalog NumberQuantity
A352201 Bundle
Catalog number A35220
Price (MXN)
-
Quantity:
1 Bundle
Automate your flow cytometry workstation with the Attune NxT and Orbitor RS2 Automation Bundle. The Orbitor RS2 Microplate Mover is an automation robot that delivers improved walk-away time and smarter productivity. This Automation Bundle is designed for an Attune NxT Acoustic Focusing Cytometer integrated with the Attune NxT, CytKick, or CytKick Max autosampler. To address your plate format storage and handling, there are three different configurations: one with three sequential access stacks (40 plates per stack) (Cat. No. A33007), one with three random access hotels (24 plates/3 hotels) (Cat. No. A33008), and one with two sequential access stacks and one random access hotel (this page).
Extensive vertical reach allows the Orbitor RS2 Microplate Mover to access virtually any microplate instrument location, including stacked instruments, and its telescoping arm can reach a variety of distances. The ingenious arm travels through the base, resulting for rapid and efficient movements. This robot has a full 360°workspace with unlimited base rotation; this eliminates the need for hard stops and reverse movements.
Advanced closed-loop motion control provides collision detection and position monitoring, allowing the Orbitor RS2 Microplate Mover to quickly resume without the need to re-home. With a wide range of grip-control functionality, this robot combines performance with innovation. From variable force control to plate detection, the feature set is designed to impress. The result is remarkable speed, precise motion, and consistent, accurate plate placement.
The Orbitor RS2 Microplate Mover is powered by Thermo Scientific Momentum Integration Software with features such as Momentum scheduling software, error detection and recovery, and a dynamic scheduler that allows changes while running.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Block FormatNon-Interchangeable
Product LineAttune
Quantity1 Bundle
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune NxT Acoustic Focusing Cytometer
Product TypeAutomation Bundle
Unit SizeEach
Contents & Storage
• 1 Orbitor RS2 Microplate Mover
• Integrated barcode reader
• 1 Momentuum Software License
• 2 plate stacks
• 1 plate hotel