Can I purchase the sample block for the SimpliAmp Thermal Cycler (Cat. No. A24811) as a standalone product? Is it replaceable/interchangeable?
Unfortunately, we do not sell the sample block for the SimpliAmp Thermal Cycler separately. Only the ProFlex series and the Automated series of thermal cyclers have interchangeable blocks that we sell separately.