TrueGuide™ gRNA - FAQs
TrueGuide™ gRNA - FAQs
View additional product information for TrueGuide™ gRNA - FAQs (A35510)
1 product FAQs found
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?
The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.