Product Image
Invitrogen™
TrueGuide™ sgRNA Positive Control, CDK4 (human)
Thermo Fisher Scientific provides a complete collection of validated positive and negative controls for the TrueGuide Synthetic gRNA product lineRead more
Have Questions?
Catalog NumberQuantity
A355233 nmol
Catalog number A35523
Price (USD)
203.00
Each
Quantity:
3 nmol
Price (USD)
203.00
Each
Thermo Fisher Scientific provides a complete collection of validated positive and negative controls for the TrueGuide Synthetic gRNA product line to help maximize your editing performance. This positive control against CDK4 targets a region of Exon 2 and has achieved editing efficiencies of up to 80% in a variety of human cell lines. Use the CDK4 control to easily determine the optimal conditions for delivering CRISPR/Cas9 tools to your cell type, allowing you to move forward in your editing experiments with confidence and minimize downstream effort to isolate and validate your edited clones.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineTrueGuide
Product TypegRNA
Quantity3 nmol
Shipping ConditionRoom Temperature
Control TypePositive Control
FormatTube
RNAi TypegRNA
SpeciesHuman
Unit SizeEach
Contents & Storage
Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
Frequently asked questions (FAQs)
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?
The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.