Cathode Buffer Container Reservoir Septa for SeqStudio™ Genetic Analyzer - FAQs
Cathode Buffer Container Reservoir Septa for SeqStudio™ Genetic Analyzer - FAQs
View additional product information for Cathode Buffer Container Reservoir Septa for SeqStudio™ Genetic Analyzer - FAQs (A35640)
2 product FAQs found
What does the SeqStudio Starter Kit (PN A35000) contain?
The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
How many times can I reuse the Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer?
The Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer are intended for single use only. We recommend replacing the Cathode Buffer Container Reservoir Septa after each run. If rinsed with water and reused, the septa can become sticky and gummy over time and may eventually start sticking to the array. This may potentially increase the risk of damaging the array.
Bleach, detergents, or solvents should not be used to clean the septa as this can cause the septa to dry out faster and risk contamination of the system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.