Search
View additional product information for SeqStudio™ Genetic Analyzer System with SmartStart - FAQs (A35644, A35645, A35646)
3 product FAQs found
SeqStudio Genetic Analyzer has not been tested for operation in low humidity conditions and is validated for environments with 20-80% humidity. Therefore, we do not recommend using SeqStudio Genetic Analyzer in below 20% humidity.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
No, since the SeqStudio instrument's data collection software is on the instrument itself, it is not necessary that you upgrade it.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.
- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.