What does the special character ˆ indicate in the library format for MicroSeq ID 16S rDNA 500 Supplemental Library?
The special character indicates that the sequence was derived from the type strain.
Search
View additional product information for MicroSEQ™ ID 16S rDNA 500 Supplemental Library - FAQs (A35993)
6 product FAQs found