GlycanAssure™ AutoXpress Script Card
Applied Biosystems™
GlycanAssure™ AutoXpress Script Card
The GlycanAssure AutoXpress Script Card is included with the Automate Express System for GlycanAssure AutoXpress Kits, which enables the hands-freeRead more
Have Questions?
Catalog NumberQuantity
A361641 card
Catalog number A36164
Price (USD)
373.00
Each
Quantity:
1 card
Price (USD)
373.00
Each
The GlycanAssure AutoXpress Script Card is included with the Automate Express System for GlycanAssure AutoXpress Kits, which enables the hands-free automation of the preparation of labelled glycans from glycoprotein samples. The script card provides the code that controls the movements of the Automate Express instrument as it moves liquids among the pre-filled cartridges until a final sample elute is achieved. Current owners of an Automate Express Nucleic Acid Extraction System who are interested in the glycan workflow can order this script card along with GlycanAssure AutoXpress kits to enable that workflow on their system.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 card
For Use With (Application)Labelled Glycan Preparation, Automate Express system for GlycanAssureAutoXpress Kits
Unit SizeEach