Ion AmpliSeq ERCC Companion Panel
Ion Torrent™
Ion AmpliSeq ERCC Companion Panel
The Ion AmpliSeq RNA ERCC Companion Panel works with the ERCC RNA Spike-In Mix to provide performance assessment of IonRead more
Have Questions?
Catalog NumberQuantity
A3655296 reactions
Catalog number A36552
Price (USD)
1,578.00
Each
Quantity:
96 reactions
Price (USD)
1,578.00
Each
The Ion AmpliSeq RNA ERCC Companion Panel works with the ERCC RNA Spike-In Mix to provide performance assessment of Ion AmpliSeq Transcriptome libraries in gene expression experiments. Add the ERCC RNA Spike-In Mix to each RNA sample, then amplify targets with the Ion AmpliSeq Transcriptome panel plus the Ion AmpliSeq RNA ERCC Companion Panel during library preparation. When analyzing the sequencing results, the data from sequenced ERCC targets is compared to the known concentration of the ERCC targets in the Spike-In Mix.
Working with the ERCC RNA Spike-In Mix, the Ion AmpliSeq RNA ERCC Companion Panel offers:
• Standard baseline of Ion AmpliSeq Transcriptome gene expression experiments across multiple experiments and various platforms
• Measurement of the lower limit of detection and dynamic range of an expression experiment
• Identification of degraded or low-yield samples by assessing the performance of ERCC targets
Variation in RNA expression data can be attributed to a variety of factors including the quality of the starting material, the level of cellularity and RNA yield, the platform employed, and the person performing the experiment. To control for these sources of variability, a common set of external RNA controls has been developed by the External RNA Controls Consortium (ERCC), an ad-hoc group of academic, private, and public organizations hosted by the National Institute of Standards and Technology (NIST). The controls consist of a set of unlabeled, polyadenylated transcripts designed to be added to an RNA analysis experiment after sample isolation, in order to measure against defined performance criteria.
The Ion AmpliSeq RNA ERCC Companion Panel consists of 10 primer pairs targeting 10 selected transcripts from the ERCC RNA Spike-In Mix representing a 13log2 dynamic range. Chosen for general GC balance and amenability for linear amplification (TaqMan qPCR), the selected ERCC targets produce R2 >0.9 under normal experimental conditions.
Specifications
For Use With (Application)RNA Sequencing
For Use With (Equipment)Ion GeneStudio™ S5 System
Product TypeERCC Companion Panel
Quantity96 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineIon AmpliSeq
Starting MaterialRNA
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
How many reactions can I perform with the Ion AmpliSeq RNA ERCC Companion Panel?
The Ion AmpliSeq RNA ERCC Companion Panel provides enough reagents to perform 96 manual reactions or 48 automated reactions.
What software do I need to view data from the Ion AmpliSeq RNA ERCC Companion Panel?
To view data from the Ion AmpliSeq RNA ERCC Companion Panel, Torrent Suite Software version 5.10 or later is required and Version 5.10.0.3 or later of the ERCC_Analysis plugin for Torrent Suite Software is required.
Can the Ion AmpliSeq RNA ERCC Companion Panel be used with FFPE samples?
We do not recommend using the Ion AmpliSeq RNA ERCC Companion Panel with FFPE samples or with any panels besides human and mouse Ion AmpliSeq transcriptome gene expression panels.
How much of the Ion Ampliseq RNA ERCC Companion Panel should I add to the target amplification reactions?
We recommend adding 1 µL of the Ion Ampliseq RNA ERCC Companion Panel per manual target amplification reaction and 8 µL per primer pool for an automated target amplification reaction on the Ion Chef Instrument.
Why would one use the Ion AmpliSeq ERCC Companion Panel?
The Ion AmpliSeq ERCC Companion Panel is compatible with Ion AmpliSeq Transcriptome Human Gene Expression Kit and Ion AmpliSeq Transcriptome Mouse Gene Expression Kit (not recommended for use with FFPE samples or panels outside of Ion AmpliSeq Human or Mouse Transcriptome kits). With the addition of the Ion AmpliSeq RNA ERCC Companion panel to your Ion AmpliSeq RNA panel, dynamic range and sensitivity can be similarly evaluated in Ion AmpliSeq RNA libraries that are prepared from RNA samples containing the ERCC RNA Spike-In Mix