MagMAX™ DNA Multi-Sample Ultra 2.0 Wash I Solution
Applied Biosystems™

MagMAX™ DNA Multi-Sample Ultra 2.0 Wash I Solution

This Wash I solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. It is available separately for applicationsRead more
Have Questions?
Catalog NumberQuantity
A365801.1 L
Catalog number A36580
Price (USD)
876.65
Online exclusive
974.00
Save 97.35
Each
Quantity:
1.1 L
Price (USD)
876.65
Online exclusive
974.00
Save 97.35
Each
This Wash I solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. It is available separately for applications that require more Wash Isolution than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineMagMAX
Product TypeDNA Multi-Sample Ultra 2.0 Wash I Solution
Quantity1.1 L
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Are there larger volume standalone reagent components available to scale up the MagMAX HMW DNA workflow?

Yes, the following larger volume standalone components are available for purchase separately:
- MagMAX HMW DNA Booster, 1,000 preps (1.0 mL) (Cat. No. A36625)
- MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K (45 mL) (Cat. No. A36578)
- MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (45 mL) (Cat. No. A36579)
- MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution (1.1 L) (Cat. No. A36580)
- MagMAX DNA Multi-Sample Ultra 2.0 Lysis Binding Solution (450 mL) (Cat No. A36581)
- MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution (120 mL) (Cat No. A36582)
- MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution (45 mL) (Cat No. A36583)
- MagMAX Cell and Tissue DNA Extraction Buffer (600 mL) (Cat No. A45470)

If I have already purchased the MagMAX DNA Multi-Sample Ultra 2.0 Kit or standalone components, what do I need to extract high molecular weight DNA?

In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or
In addition to the MagMAX DNA Mult-Sample Ultra 2.0 product(s) you will need:
MagMAX HMW DNA Booster, 100 preps (Cat. No. A36624) or MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).
MagMAX HMW DNA Booster, 1,000 preps (Cat No. A36625).

For cell and tissue samples include:
MagMAX Cell and Tissue DNA Extraction Buffer, 100 preps (Cat. No. A45469) or MagMAX Cell and Tissue DNA Extraction Buffer, 1,000 preps (Cat. No. A45470)
The MagMAX HMW DNA User Guide (MAN1001478)
The specific HMW DNA KingFisher automation protocol for your KingFisher Purification System (see product description page for download).

Can the MagMAX HMW DNA Booster be paired with other genomic DNA isolation kits?

The MagMAX HMW DNA Booster is specifically designed and optimized for use with the MagMAX DNA Multi-Sample Ultra 2.0 (Cat. No. A36570) chemistry.