MicroAmp™ EnduraPlate™ Optical 96-Well Fast Clear Reaction Plates
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Fast Clear Reaction Plates

MicroAmp EnduraPlate Optical 96-Well Fast Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs. EnduraPlateRead more
Have Questions?
Catalog NumberQuantity
A3693020 plates
Catalog number A36930
Price (MXN)
-
Quantity:
20 plates
MicroAmp EnduraPlate Optical 96-Well Fast Clear Reaction Plates offer excellent PCR or qPCR performance to meet your experimental needs. EnduraPlate plastic consumables are validated with Applied Biosystems instruments for optimal fit and performance. They are constructed to ANSI/SBS standards and are well-suited for automated workflows. MicroAmp EnduraPlate Optical 96-Well Fast Clear Reaction Plates are semi-skirted and fit low-profile/Fast (0.1 mL) 96-well thermal blocks.

• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNase-, and PCR inhibitor-free
• Constructed to ANSI/SBS standards for compatibility with high-throughput laboratories and robotic handling
• Easy-to-read black well-identification text to minimize pipetting errors

EnduraPlate reaction plates use polypropylene for a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from the thermal block to the sample, while a polycarbonate frame provides mechanical stability, flatness, and durability that is compatible with automation/robotics. Easy-to-read, black well-identification text helps to minimize pipetting mistakes and improves sample preparation.

MicroAmp EnduraPlate Optical 96-Well Fast Clear Reaction Plates are provided without barcodes, enabling customization with your own barcodes. For a barcoded version of this product, please see MicroAmp EnduraPlate Optical 96-Well Fast Clear Reaction Plates with Barcode.

Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Packaging20 Plates
Product TypeMicroplate
Quantity20 plates
Shipping ConditionRoom Temperature
Skirt StyleSemi
Volume (Metric) Working0.1 mL
BarcodeNo Barcode
ColorOptical, Clear
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineMicroAmp
Volume (Metric) Well0.2 mL
Unit SizeEach
Contents & Storage
Store at room temperature.