Oncomine™ Myeloid Research Assay
Ion Torrent™

Oncomine™ Myeloid Research Assay

The Ion Torrent Oncomine Myeloid Research Assay consists of three pools of AmpliSeq oligonucleotide primers and associated reagents to generateRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Samples
A36940Up to 24 Samples
A51770Up to 96 Samples
Catalog number A36940
Price (USD)
12,010.00
Each
No. of Samples:
Up to 24 Samples
Price (USD)
12,010.00
Each

The Ion Torrent Oncomine Myeloid Research Assay consists of three pools of AmpliSeq oligonucleotide primers and associated reagents to generate amplicon libraries for next-generation sequencing (NGS) on Ion Torrent platforms. The assay is designed to provide sensitive and comprehensive sample amplification of relevant DNA mutations and fusion transcripts associated with myeloid disorders in a single NGS run. The panel is comprised of 40 key DNA genes and a broad fusion panel of 29 driver genes to cover the most relevant targets in major myeloid disorders: acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML).

This version of the assay is for manual library preparation. For automated library preparation using the Ion Chef System, please see Cat. No. A36941.

Features include:

  • Simple, robust workflow that encompasses mutations and fusions in two DNA and one RNA pool in a single NGS run
  • Excellent coverage of challenging targets such as CEBPA and FLT3-ITDs
  • Multiplex up to four samples on a single Ion 318 chip or twelve on an Ion 530 chip
  • Compatible with as little as 10 ng/pool input DNA or RNA per library from blood or bone marrow samples
  • Validated detection of somatic variants down to 5% allele frequency
  • Simple and fast workflow produces targeted libraries in 3.5 hours typically
  • For use on the Ion PGM and Ion S5 systems with manual or automated library preparation
  • Compatible with the Oncomine Knowledgebase Reporter for variant annotation

The Oncomine Myeloid Research Assay is a comprehensive, targeted NGS assay designed to assist hematologists in the understanding of myeloid cancer. The assay is a complete kit facilitating the amplification of multiple gene targets from blood or bone marrow samples with as little as 10 ng of input DNA or RNA per pool. Leveraging the power of Ion AmpliSeq technology, this highly multiplexed NGS assay enables the generation of results from multiple samples in a single run. Designed for use on either the Ion PGM or Ion S5 sequencing systems, results are delivered in days rather than weeks. The assay is aligned with bioinformatics workflows within Torrent Suite and Ion Reporter analysis software that utilize optimized variant-calling parameters for SNV, InDel, and FLT3 large internal tandem duplication (ITD) detection. Variants can be annotated in the Oncomine Knowledgebase Reporter to generate a sample-specific report. Samples can be processed quickly and easily, and variants detected and identified confidently.

Specifications
For Use With (Application)Oncology Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of Reactions24
No. of SamplesUp to 24 Samples
Nucleic Acid TypeDNA and RNA
Product LineOncomine
Product TypeAssay
Quantity24 reactions
Sample TypeWhole Blood, Bone Marrow
Shipping ConditionDry Ice
ThroughputUp to 12 Samples per Ion 530 Chip
Workflow StepPanel and Library
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

What extraction kit is compatible with the Oncomine Myeloid Research Assay (Cat. Nos. A36940, A36941)?

We have validated the use of the MagMAX Total Nucleic Acid Isolation Kit (Cat. No. AM1840), which is compatible with whole blood and bone marrow samples. We recommend using your preferred commercially available kit to sequentially isolate high quality gDNA and RNA from research samples for use in library preparation.

What are the sample input requirements for the Oncomine Myeloid Research Assay (Cat. No. A36940)?

10 ng of gDNA per pool at a minimum concentration of ≥1.48 ng/µL for manual library preparation 10 ng of RNA at a minimum concentration of ≥2.5 ng/µL for manual library preparation

What sample types can I use with the Oncomine Myeloid Research Assay (Cat. Nos. A36940, A36941)?

Blood and bone marrow samples are compatible with the Oncomine Myeloid Research Assay. This kit is not intended for use with FFPE samples.

Which templating and sequencing kits are compatible with the Oncomine Myeloid Research Assay (Cat. Nos. A36940, A36941)?

The kit has been validated on the Ion GeneStudio S5 Series Systems and Ion PGM System.

With the Ion GeneStudio S5 Series Systems, we recommend using:
-Ion 510 & Ion 520 & Ion 530 Kit - Chef (Cat. No. A34019)
-Ion 530 Chip Kit (Cat. No. A27764).

With the Ion PGM System, we recommend using:
-Ion PGM Hi‑Q View OT2 Kit (Cat. No. A29900)
-Ion PGM Hi‑Q View Chef 400 Kit (Cat. No. A30798)
-Ion PGM Hi‑Q View Sequencing Kit (Cat. No. A30044)
-Ion 318 Chip v2 BC (Cat. No. 4488150)
Note: Ion 540 Chip is NOT compatible with the Oncomine Myeloid Research Assay as 400 base pair (850 Flows) sequencing is required.

How many samples can I multiplex with the Oncomine Myeloid Research Assay (Cat. Nos. A36940, A36941)?

The assay can be run on both the Ion PGM System and Ion S5/S5XL systems.
-PGM Plexy = 4 samples per Ion 318 Chip
-S5 Manual Library Prep Plexy = 12 samples per Ion 530 Chip
-S5 Chef Library Prep Plexy = 8 samples per Ion 530 Chip