Ion 550™ Single Chip Supplemental Kit - FAQs
Ion 550™ Single Chip Supplemental Kit - FAQs
View additional product information for Ion 550™ Single Chip Supplemental Kit - FAQs (A36953)
3 product FAQs found
Is Ion S5 Sequencing Reagents (Part No. A27768) used for both the Ion 540 and Ion 550 Chip sequencing?
Yes, Ion S5 Sequencing Reagents (Part No. A27768) is used on the instrument for both Ion 540 and Ion 550 Chip sequencing.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center
Do I need to purchase the Ion 550 Single Chip Supplemental Kit?
The Ion 550 Kit-Chef contains Ion Chef templating and sequencing consumables sufficient for preparing and sequencing eight Ion 550 chips for four Ion Chef runs and four sequencer initializations (dual chip run configuration). If you are running in the single chip configuration, then you will need to purchase the Ion 550 Single Chip Supplemental Kit separately to ensure that you have enough templating and sequencing consumables to support eight single chip Ion Chef templating runs and eight sequencer initializations with the Ion 550 Kit-Chef.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is in the Ion 550 Single Chip Supplemental Kit and how is it shipped/stored?
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required to support eight single chip Ion Chef runs and eight sequencer initializations with the Ion 550 Kit-Chef. The Ion 550 Single Chip Supplemental Kit includes the following components:
- Ion S5 Chef Supplies (4 boxes) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Solutions (1 box) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Reagents (1box) (ships on wet ice at 2-8 degrees C; store at -20 degrees C)
IMPORTANT! Do not store the Ion S5 Sequencing Reagents (Part No. A27768) on dry ice or in a closed environment containing dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.