What are the characteristics of BT-549 Control Lysate, and what is its main purpose?
BT-549 Control Lysate is a stimulated BT-549 cell lysate that has been validated for the expression levels and phosphorylation of key targets (TP53 and phosphorylated TP53) in the TP53 signaling pathway. It serves as a positive control for use with SureQuant TP53 panel kits.