How can I sign in as an administrator to the MiniAmp Plus Thermal Cycler (Cat. No. A37835, A37835, A38076, A38077) and connect to the cloud? What is the default admin password?
The default admin password/PIN is 0000 + the last 4 digits of the instrument serial number. For example, if the serial number was 1101234567, then the default admin password would be 00004567.
Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.
Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.