Ion AmpliSeq™ Exome RDY Kit 1x8 - FAQs
Ion AmpliSeq™ Exome RDY Kit 1x8 - FAQs
View additional product information for Ion AmpliSeq™ Exome RDY Kit 1x8 - FAQs (A38262)
4 product FAQs found
What is the shipping condition for Ion AmpliSeq Exome RDY Kit 1x8 (Cat. No. A38262)?
The kit is shipped on wet or dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many runs is Ion AmpliSeq Exome RDY Kit 1x8 (Cat. No. A38262) good for?
The kit contains reagents for preparation of 8 exome libraries for sequencing a plate containing 12 primer pools that are dried into 8 rows.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for the Ion AmpliSeq Exome RDY kits?
We recommend storing at 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the components of Ion AmpliSeq Exome RDY Kit, 1x8 (Cat. No. A38262)?
Here are the kit components:
- Ion AmpliSeq Exome RDY Panel 1x8 (1 plate for 8 libraries)
- Ion AmpliSeq Library Kit Plus (sufficient for preparing eight exome libraries)
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.