Ion AmpliSeq™ Exome RDY Kit 4x2 - FAQs
Ion AmpliSeq™ Exome RDY Kit 4x2 - FAQs
View additional product information for Ion AmpliSeq™ Exome RDY Kit 4x2 - FAQs (A38264)
5 product FAQs found
What is the shipping condition for Ion AmpliSeq Exome RDY Kit 4x2 (Cat. No. A38264)?
The kit is shipped on wet or dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many runs is Ion AmpliSeq Exome RDY Kit 4x2 (Cat. No. A38264) good for?
The kit contains reagents for preparation of 8 exome libraries for sequencing plates containing 12 primer pools that are dried into 2 rows of 4 plates
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion AmpliSeq Exome RDY Kit 4x2 (Cat. No. A38264)?
We recommend storing at 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the components of Ion AmpliSeq Exome RDY Kit 4x2 (Cat. No. A38264)?
Here are the kit components:
- Ion AmpliSeq Library Kit Plus (Part No. 4488990), qty. 1
- Ion AmpliSeq Exome RDY Panel 4x2 (Part No. 4489840), 4 plates for 2 libraries each
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for the Ion AmpliSeq Exome RDY kits?
We recommend storing at 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.