7(Z),11(Z)-Pentacosadiene (CAY-9000531-10mg) - FAQs
7(Z),11(Z)-Pentacosadiene (CAY-9000531-10mg) - FAQs
View additional product information for 7(Z),11(Z)-Pentacosadiene (CAY-9000531-10mg) - FAQs (A38402)
2 product FAQs found
What is the shelf life for One Shot FBS?
The current shelf life for One Shot FBS products is 2 years from the date of manufacture.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the recommended storage temperature for your FBS products?
We recommend storing our FBS products at ≤-10 degrees C. The current United States Pharmacopeia (USP) and European Pharmacopoeia (EP) guidelines for monographs for serum refer to a storage temperature of ≤-10 degrees C.
While our previous recommended storage temperature of -5 to -20 degrees C have complied with this range, the change to ≤-10 degrees C ensures that the storage temperature range on the label and CoA is consistent with USP and EP requirements.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.