Product A38447 is equivalent software to Cat. No. A30371.
Product Image
Applied Biosystems™

MicroSEQ™ ID Microbial Identification Software v3.1 Bundle

MicroSEQ ID Microbial Identification Software v3.1 allows you to easily identify and classify unidentified bacterial or fungal sequences by comparingRead more
Have Questions?
Catalog NumberQuantity
A384471 CD
Catalog number A38447
Price (USD)
71,400.00
Each
Quantity:
1 CD
Price (USD)
71,400.00
Each
MicroSEQ ID Microbial Identification Software v3.1 allows you to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library. This bundle includes the bacterial and fungal sequence libraries.

• A validated library of over 3000 microbial sequences helps ensure you get the answers you need
• Automatically identify unknown specimens
• Run plates on Applied Biosystems 3500 Series genetic analyzers directly via the MicroSEQ ID software
• The MicroSEQ system is a complete integrated solution, including security, audit, and e-signature features
• Qualification and validation support are available

Confidently identify microbes
The MicroSEQ ID sequence libraries consist of bacterial libraries of 500 bp 16S rDNA sequences and approximately 9300 validated entries, including extensive coverage of gram-negative non-fermenters, Bacillus, Coryneforms, Mycobacteria, and Staphylococcus, as well as a fungal library containing more than 1300 entries. To help ensure accurate identification of the species of your sequences, we rigorously validate all library entries. And unlike other widely available databases, MicroSEQ libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections. The libraries also support sequence polymorphisms, and include bacterial Category A biothreat agents.

Automatically identify unknown specimens
Reviewing each specimen in a project file to make a manual identification call can be tedious and time consuming. The Auto-ID feature in MicroSEQ ID software uses a set of editable parameters to automatically identify unknown specimens. The results table is displayed in the report, and an editable comments field allows additional information or manual identifications to be entered into the same table.

Run plates on 3500/3500xL instruments directly via the MicroSEQ ID software
Applied Biosystems 3500 Series genetic analyzer users are now able to run plates on these instruments directly via the MicroSEQ ID software v3.1 user interface, without needing Autoanalysis Manager. A single software wizard enables you to set up projects, specimens, and plates; adjust the plate layout via drag and drop; start and monitor the run; and review results.

The MicroSEQ system: A complete, integrated solution
Streamlining every step in microbial identification, the MicroSEQ system combines the advantages of MicroSEQ ID software with easy-to-use PrepMan Ultra sample preparation reagents and protocols, MicroSEQ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To help ensure optimal performance and system-wide integration, we develop and test all components of the system together. The MicroSEQ ID software also includes security, audit, and electronic signature features enabling the software to smoothly integrate into your workflow.

Qualification and validation support
To help ensure seamless integration into your laboratory, from installation to routine operation, we offer comprehensive installation and operational qualification (IQ/OQ) and validation support services.

Specifications
For Use With (Application)Sequencing
Product LineMicroSEQ
Product TypeID Microbial Identification Software
Quantity1 CD
Shipping ConditionRoom Temperature
SoftwareSequence Analysis, Identification
Unit SizeEach
Contents & Storage
• MicroSEQ ID software version 3.1 (Full)
• 500 16S rDNA bacterial library
• 500 16S rDNA supplemental bacterial library
• 16S Full Gene bacterial library
• D2 Fungal library

Frequently asked questions (FAQs)

What is included in the MicroSEQ ID Microbial Identification Software v3.1 Bundle, Cat. No. A38447?

The components of the MicroSEQ ID Microbial Identification Software v3.1 Bundle (Cat. No. A38447) are the following:
- MicroSEQ ID software version 3.1 (Full)
- 500 16S rDNA bacterial library
- 500 16S rDNA supplemental bacterial library
- 16S Full Gene bacterial library
- D2 Fungal library

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

You offer two MicroSEQ ID Microbial Identification Software v3.1 Bundle kits, Cat. Nos. A30371 and A38447? What is the difference between them?

MicroSEQ ID Microbial Identification Software v3.1 Bundle, Cat. No. A38447, includes all the components of Cat. No. A30371 and in addition, it includes the validated 500 16S rDNA supplemental bacterial library. Please note that Cat. No. A38447 will be replacing Cat. No. A30371.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.