Pro-Detect™ Rapid Human Fc Assay Kit
Thermo Scientific™

Pro-Detect™ Rapid Human Fc Assay Kit

The Thermo Scientific Pro-Detect Rapid Human Fc Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detectionRead more
Have Questions?
Catalog NumberQuantity
A3850410 Assays
Catalog number A38504
Price (USD)
152.00
Each
Quantity:
10 Assays
Price (USD)
152.00
Each
The Thermo Scientific Pro-Detect Rapid Human Fc Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of human Fc-tagged proteins in protein expression and purification systems. This lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the human Fc tag to provide a visual, color readout for detection within ten minutes. This assay can be used to detect human Fc-tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.

Features of the Pro-Detect Rapid assay kits:
• Easy to adopt—lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
• Simple, fast method—simply dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5-10 minutes
• Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
• Specific—assay does not cross-react with common lysis and protein preparation reagents
• Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
• Long shelf life—stable for at least one year at 4°C

Each Pro-Detect Rapid assay is performed by simply adding a properly diluted tissue culture supernatant, lysate, or purified protein sample to a test tube or microplate well and dipping in a lateral flow strip. Gold-conjugated detection antibodies embedded in the strip form complexes with the tagged protein in the sample. These complexes travel the length of the strip membrane and bind at the test and control lines. Results are displayed as red bands.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayLateral Flow Rapid Assay
Detectable AnalytesHuman Fc-tagged proteins
For Use With (Application)Lateral Flow Rapid Assay
Product LinePro-Detect
Product TypeAssay
Quantity10 Assays
Shipping ConditionWet Ice
SpecificityAntibody
Sufficient For10 assays
Detection MethodChromogenic
Sample TypeCell Culture Supernatant, Cell Lysates
Target SpeciesHuman
Unit SizeEach
Contents & Storage
• 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.

Frequently asked questions (FAQs)

With Ion S5 2X Cleaning Solution, how long does a "pulse clean" take to complete?

It takes approximately 47 mins to do a "pulse clean",

When using Ion S5 2X Cleaning Solution, where in the software should I select "pulse clean"?

Go to "System tools" and select "pulse clean". The instrument must already have been cleaned after a run.

What are the dimensions of the bottles in Ion S5 2X Cleaning Solution?

Here are the bottle dimensions:
- Length (Metric) 61 mm
- Height (Metric) 131.1 mm
- Width (Metric) 61 mm

How many bottles of Ion S5 2X Cleaning Solution do you provide in Cat. No. A34172?

The kit contains 2 bottles of Ion S5 2X Cleaning Solution.

Which instruments is Ion S5 2X Cleaning Solution compatible with?

Ion S5 2X Cleaning Solution is compatible with GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems.