Inquire about OEM or Commercial Supply version of this product here.
Collibri™ PCR-free PS DNA Library Prep Kits for Illumina Systems
Invitrogen™

Collibri™ PCR-free PS DNA Library Prep Kits for Illumina Systems

The Invitrogen Collibri PCR-free PS DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes, including FFPE samples.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수제품 유형
A3860902424 PrepsPCR-free PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
A38608024W24 PrepsPCR-free PS DNA Library Prep Core Kit
A38610096W96 PrepsPCR-free PS DNA Library Prep Core Kit
A3860802424 PrepsPCR-free PS DNA Library Prep Kit with CD Indexes
A3861009696 PrepsPCR-free PS DNA Library Prep Kit with CD Indexes
A4360802424 PrepsPCR-free PS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
A4360902424 PrepsPCR-free PS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
A4361002424 PrepsPCR-free PS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
A3861519696 PrepsPCR-free PS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
카탈로그 번호 A38609024
제품 가격(KRW)
1,432,000
온라인 행사
Ends: 31-Dec-2026
1,684,000
할인액 252,000 (15%)
Each
반응 수:
24 Preps
제품 유형:
PCR-free PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
1,432,000
온라인 행사
Ends: 31-Dec-2026
1,684,000
할인액 252,000 (15%)
Each
The Collibri PCR-free PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from inputs as low as 500 ng of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PCR-free PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from inputs as low as 500 ng of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PCR-free PS DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri PCR-free PS DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. The kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification and sequencing on Illumina instruments.

Applications
The Collibri PCR-free PS DNA Library Preparation Kit is recommended for:
• Whole genome sequencing
• Resequencing
• De novo assembly

For Research Use Only. Not for use in diagnostic procedures.
사양
설명The product contains modules A38576024, A38587024, A43190024
용도(애플리케이션)Whole Genome Sequencing, Resequencing, De Novo Assembly
용도(장비)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
라이브러리Fragment Library
반응 수24 Preps
제품라인Collibri
제품 유형PCR-free PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
수량24 preps
샘플 종류DNA
배송 조건Dry Ice
종All
워크플로우 단계Library Generation
형식Kit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
구성 및 보관
• Collibri PCR-free PS DNA Library Prep Kit, store at -20°C
• Collibri DNA Library Cleanup Kit, store at 4°C
• Collibri DNA UD Indexes, store at -20°C

자주 묻는 질문(FAQ)

Can indexed adaptors or other modules be purchased separately for the Collibri DNA Library Prep kits?

Indexed adapters and other modules in the kit can be purchased separately. Please refer to thermofisher.com/collibriDNA and contact us for more details.

With the Collibri DNA Library Prep kits, what is the distribution of enzymatically sheared DNA in the library?

Fragment size ranges from 150 bp to 850 bp. For optimal results, we recommend and provide detailed instructions for 350 bp and 550 bp average size library construction.

With the Collibri DNA Library Prep kits, can I use indexed adaptors interchangeably between RNA-seq and DNA-seq applications?

No. They are specifically designed for a product and cannot be used interchangeably.

With the Collibri DNA Library Prep kits, why is my ligation master mix orange instead of red?

Dyes inside the master mix react with CO2 and change the color from red to orange. The mix is still usable when the color changes to orange. To prevent discoloration, avoid keeping it on dry ice.

Are Collibri DNA Library Prep kits compatible with FFPE samples?

Collibri PS DNA Library Prep Kit can be used for library construction from lower-quality DNA (such as FFPE samples). However, the yields of adaptor-ligated molecules are typically lower compared to the yields obtained with high-quality DNA.

Collibri ES DNA Library Prep Kit is not recommended for library construction from FFPE samples.

인용 및 참조 문헌 (2)

인용 및 참조 문헌
Abstract
Targeted In Situ Protein Diversification and Intra-organelle Validation in Mammalian Cells.
Authors:Erdogan M, Fabritius A, Basquin J, Griesbeck O
Journal:Cell Chem Biol
PubMed ID:32142629
Engineered proteins must be phenotypically selected for function in the appropriate physiological context. Here, we present a versatile approach that allows generating panels of mammalian cells that express diversified heterologous protein libraries in the cytosol or subcellular compartments under stable conditions and in a single-variant-per-cell manner. To this end we adapt ... More
Effects of Prenatal Phthalate Exposure and Childhood Exercise on Maternal Behaviors in Female Rats at Postpartum: A Role of
Authors:Lee YJ, Lin HT, Chaudhary MA, Lee YC, Wang DC
Journal:Int J Mol Sci
PubMed ID:34576011
Both the detrimental effect of prenatal exposure to di-(2-ethylhexyl)-phthalate (DEHP) and the beneficial effects of physical exercise on brain functions have been reported. The oxytocin pathway has been implicated in the onset of maternal behaviors. Epigenetic modification of the oxytocin receptor gene (OXTR) through DNA methylation has been associated with ... More