Product Image
Applied Biosystems™
GeneMapper™ Software 5, client installation
Note: This client version of GeneMapper™ Software 5 installs the client configuration (user interface) only. Users must install GeneMapper™ SoftwareMás información
Have Questions?
Número de catálogoCantidad
A388921 licencia
Número de catálogo A38892
Precio (MXN)
-
Cantidad:
1 licencia
Note: This client version of GeneMapper™ Software 5 installs the client configuration (user interface) only. Users must install GeneMapper™ Software 5, full installation, prior to installing the client version, which should be installed on a computer separate from that of the full version.
GeneMapper™ Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all of our electrophoresis-based genotyping systems. This software specializes in multi-application functionality, including amplified fragment length polymorphism (AFLP™), loss of heterozygosity (LOH), microsatellite and SNP genotyping analysis. GeneMapper™ Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment. The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping. In addition, the security and audit features help users meet 21 CFR 11 requirements.
Features include:
• Installation on the data collection computer or a non-instrument computer
• Support of networked multiple clients when installed on a non-instrument computer
• An Oracle™ 11g database with project-related data storage that ensures faster installation times and latest database support, as well as flexible archive capabilities
• Customizable project auto-saving frequency
• Defining of linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800bps)
• Analysis of 6-dye data generated from the Applied Biosystems™ 3500 and 3500xL instrument systems for higher multiplexing capabilities
GeneMapper™ Software has not undergone specific developmental validation for human identification applications. Human identification laboratories analyzing single-source or parentage samples that choose to use GeneMapper™ Software for data analysis should perform their own developmental validation studies.
Several users may access the database concurrently; however, because genotype analysis is data-intensive, more than five clients simultaneously accessing the database may slow the performance of GeneMapper™ Software.
Recommended Computer Configuration: Dell™ Optiplex XE, with Core 2 Duo E8400/3.0GHz, 6MB, 1333FSB, 4GB, Non-ECC, 1333MHz DDR3, 2X2GB, Dell OptiPlex 580, 2 x 500GB SATA 3.0Gb/s and 8MB Data Burst Cache, 16X DVD+/-RW SATA, Data Only, 17-inch Color Monitor (1280 x 1024 pixels or higher). Supported Operating Systems: Windows™ 7 Professional Service Pack 1 32-bit.
Especificaciones
Introducción de datosCE Fragment Analysis (.fsa files)
LicenciaClient
Sistema operativoWindows 7 Pro Service Pack 1
Tipo de productoGeneMapper Software 5 Client Installation
Cantidad1 licencia
Categoría de softwareAdmite archivos de todos los analizadores genéticos AB
Línea de productosGeneMapper
Software TypeCE Fragment Analysis
Unit SizeEach
Contenido y almacenamiento
Software/CD
Preguntas frecuentes
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.
Can the GM4 license (GeneMapper Software v4.0, client installation, Cat. No. 4440914) be used to register for GM5 (GeneMapper Software 5, client installation, Cat. No. A38892) or GM6 (GeneMapper Software 6, client installation, Cat. No. 4475074) copies?
No. You must get a GM5/GM6 license separately.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which instruments does GeneMapper Software 6 support?
GeneMapper Software 6 fully supports data generated from the following instruments running all versions of Data Collection Software:
- Applied Biosystems SeqStudio Genetic Analyzer
- Applied Biosystems 3500 Genetic Analyzer
- Applied Biosystems 3500xL Genetic Analyzer
- Applied Biosystems 3130 Genetic Analyzer
- Applied Biosystems 3130xl Genetic Analyzer
- Applied Biosystems 3730 DNA Analyzer
- Applied Biosystems 3730xl DNA Analyzer
- Applied Biosystems 310 Genetic Analyzer
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What has been updated in GeneMapper Software 6?
The primary updates in GeneMapper Software 6 are the following:
- GeneMapper Software 6 supports installation on Microsoft Windows 7 Professional and Microsoft Windows 10 Professional (64-bit) operating systems.
- GeneMapper Software 6 allows only administrative users to use Delete All Audit Records option. For non-admin users, this option is disabled.
- GeneMapper Software 6 has an enhanced Sample Information View to show consumables information.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
We just upgraded to GeneMapper Software v6, and I'm wondering if my old data files will work on it.
GeneMapper Software v6 is compatible with the following files:
-3130 series, Data Collection Software v3.0, 3.1, 4
-3730 series, Data Collection Software v3.x, 4.x
-3500 series, Data Collection Software v1.x, 2, 3.x
-SeqStudio Data Collection Software v1.x
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.