MicroSEQ™ ID Microbial Identification Software Lite v3.1 Bundle - FAQs
MicroSEQ™ ID Microbial Identification Software Lite v3.1 Bundle - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software Lite v3.1 Bundle - FAQs (A38912)
2 product FAQs found
What is included in the MicroSEQ ID Microbial Identification Software Lite v3.1 Bundle (Cat. No. A38912)?
The components of the MicroSEQ ID Microbial Identification Software Lite v3.1 Bundle (Cat. No. A38912) are the following:
- MicroSEQ ID software version 3.1 (Lite)
- 500 16S rDNA bacterial library
- 500 16S rDNA supplemental bacterial library
- 16S Full Gene bacterial library
- D2 Fungal library
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
You offer two MicroSEQ ID Microbial Identification Software Lite v3.1 Bundle kits, Cat. Nos. A30374 and A38912. What is the difference between them?
MicroSEQ ID Microbial Identification Software Lite v3.1 Bundle, Cat. No. A38912, includes all the components of Cat. No. A30374 and in addition, it includes the validated 500 16S rDNA supplemental bacterial library. Please note that Cat. No. A38912 will be replacing Cat. No. A30374.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.