GlycanAssure™ AutoXpress Kit with CE Module

Catalog number:  A38930

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The GlycanAssure AutoXpress Kit is designed for use on the AutoMate Express System for GlycanAssure AutoXpress Kits, for hands-free automation of the preparation of labelled glycans from glycoprotein samples. The kit includes GlycanAssure AutoXpress cartridges that contain pre-filled reagents and plastic tubes and tips to perform hands-free glycan release and labelling of up to 13 samples in one run. This version of the kit contains a CE module with additional chemistries needed for CE analysis.

The GlycanAssure AutoXpress cartridge reagents are based on the established GlycanAssure chemistry for N-glycan rapid release, labeling, and cleanup that applies a novel method for N-glycan release in a complete form. The GlycanAssure APTS N-glycan sample prep method consists of rapid deglycosylation using PNGase-F enzyme followed by magnetic bead-based glycan purification, glycan labeling with ATPS dye, and excess dye removal. The GlycanAssure AutoXpress Kit combined with the AutoMate Express System offers a single, automated N-glycan sample prep workflow for both high throughput (CE) and characterization (UHPLC) biopharma applications.

Key features of the combined kit/system include:
• Fully automated sample prep for N-glycan analysis
• Rapid results with cartridge-based reagents for glycan release and labelling
• High quality glycan data with reduced analyst error
• Labeled glycans can be analyzed on LC or CE platforms
• Small system footprint and low-cost automation platform make it adaptable in both development and QC labs
• Consistent data for easy method development and transfer across the globe

Specifications

Product Size: 52 reactions

Contents & storage

• GlycanAssure bead kit and LIZ Standard, store at 2–8°C
• AutoXpress cartridges, Landmark Red, store at –20°C
• CE Loading Buffer, store at room temperature

Documents

Manuals & protocols