Unlike Cat. No. A38994024, Cat. No. A38996024 comes with the Unique Dual Indexes (UDIs), listed in table 6, page 10 of the
Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide
Please also refer to the
Technical Note "How to prevent index hopping" for additional information.
In general, UDIs have a sample index that is specific to a given library. All nucleic acids in a sample are labeled with the same sequence tag and the resulting library is pooled with other libraries and sequenced in parallel in a single run. UDIs are the preferred method when multiplexing to mitigate sample misassignment due to index hopping and the reagent kits supply enough to perform extra cycles. UDIs require extra cycles and the reagent kits supply enough to perform those extra cycles. UDIs can be especially effective when using instruments with patterned flow cells, such as the NovaSeq 6000 system.
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.