Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems, with UD indexes, 96 rxns - FAQs
Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems, with UD indexes, 96 rxns - FAQs
View additional product information for Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems - FAQs (A39005024, A39003024, A38996096, A39003096, A38994096, A39005096, A38996024, A38994024)
5 product FAQs found
Is the elution buffer, included in the Collibri Stranded RNA Library Prep Kit for Illumina Systems, "Tris buffered" or "Tris-Tween"?
Collibri Stranded RNA Library Prep Kit for Illumina Systems elution buffer is based on Tris-HCl, with no EDTA or EGTA.
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.
Is there an RNA control for optimizing fragment length included with the Collibri Stranded RNA Library Prep Kit for Illumina Systems?
What are the recommended RNA input sample types and respective sample amounts for Collibri Stranded RNA Library Prep Kit for Illumina Systems?
What are the differences between all Collibri Stranded RNA Library Prep Kit for Illumina Systems catalog numbers?
What additional materials are required when using Collibri Stranded RNA Library Prep Kit for Illumina Systems?