TaqMan™ Urinary Tract Microbiota Amplification Control - FAQs
TaqMan™ Urinary Tract Microbiota Amplification Control - FAQs
View additional product information for TaqMan™ Urinary Tract Microbiota Amplification Control - FAQs (A39174)
4 product FAQs found
Does the TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) contain 16S rRNA and RNase P?
The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) contains a linearized plasmid harboring multi-target sequences, including the sequences of 16S rRNA and the RNase P genes. There is no separate 16S or RNase P control in this kit.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center
Which sample prep kit do you recommend when running urinary tract microbiota profiling experiments?
We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for urinary tract microbiota profiling experiments.
Do you have any positive controls for the urinary tract microbiota assays?
The TaqMan Urinary Tract Microbiota Amplification Control (Cat. No. A39174) is a linearized multi-target plasmid with target sequences for each available urinary tract microbiota profiling assay.
How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?
We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.