Pierce™ FlexMix™ Calibration Solution - FAQs
Pierce™ FlexMix™ Calibration Solution - FAQs
View additional product information for Pierce™ FlexMix™ Calibration Solution - FAQs (A39239)
3 product FAQs found
What is the difference between Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?
Both the Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers have the same 16 components and produce the same spectra. However, the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers has a different storage solution: LC-MS grade acetonitrilie 87% (v/v) and LC-MS grade water 13% (v/v). The FlexMix Calibration for Auto-Ready Mass Spectrometers is designed to be used only in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What type of instruments does the Pierce FlexMix Calibration Solution work with?
We recommend using the Pierce FlexMix Calibration Solution with Orbitrap Fusion, Orbitrap Fusion Lumos, and Orbitrap ID-X MS instruments.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What is the Pierce FlexMix Calibration Solution, and what is its main purpose?
The Pierce FlexMix Calibration Solution is a mixture of 16 highly pure, ionizable components (mass ranges: 50 to 3,000 m/z) designed for both positive and negative ionization calibration using Orbitrap Tribrid Series 3.1 Instrument Control Software (or later) for Orbitrap Fusion, Orbitrap Fusion Lumos, and Orbitrap ID-X MS instruments.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.