What is the shelf life of Pierce DTT (Dithiothreitol), No-Weigh Format (Cat. No. A39255)?
Pierce DTT (Dithiothreitol), No-Weigh Format is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.