Search
| Catalog Number | Quantity |
|---|---|
| A39485 | 1 package |
This package combines everything you need to streamline your DNA sample analysis workflow and increase productivity.
The kit package includes:
E-Gel Power Snap Electrophoresis System
The all-in-one Invitrogen E-Gel Power Snap Electrophoresis System is designed for fast and convenient E-Gel agarose gel electrophoresis, from DNA sample loading to gel image. With dry precast E-Gel agarose gel technology, you can run DNA samples in as little as 10 minutes and observe sample separation in real time. It is the only benchtop system that seamlessly integrates DNA sample separation and gel imaging into one workflow.
Features of the E-Gel Power Snap Electrophoresis system include:
E1-ClipTip Bluetooth Electronic Adjustable Tip-Spacing Multichannel Equalizer Pipette
Perform sample transfers between virtually any tube, rack, microplate, or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip equalizer pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to accomodate the desired labware format. The My Pipette Creator app enables programming of the E1-ClipTip pipette from the comfort of your PC.
For Research Use Only. Not for use in diagnostic procedures.
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E1-ClipTip Bluetooth Electronic Adjustable-Tip Spacing Multichannel Equalizer Pipette (Cat. No. 4672040BT)
• 10 E-Gel EX Agarose Gels, 2% (Cat. No. G401002)
• E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
• Gel opening knife (Cat. No. EI9010)
• ClipTip Filtered 384 Format Pipette Tips, 30 μL (Cat. No. 94420103)
Storage: E-Gel Agarose Gels should be stored strictly at room temperature.