Ponceau S Staining Solution - FAQs
Ponceau S Staining Solution - FAQs
View additional product information for Ponceau S Staining Solution - FAQs (A40000278, A40000278X3, A40000279)
1 product FAQs found
What is the best way to image Ponceau staining with an iBright Imaging System?
The recommended imaging method depends on the iBright imager model being used. For iBright FL1500 and CL1500 Imaging Systems, we recommend imaging Ponceau-stained samples with the Ponceau application that can be found in Universal mode. For the iBright CL750 Imaging System, we recommend imaging Ponceau-stained samples in Protein gel mode.
We always recommend imaging with the orange iBright System Contrast Tray (Cat. No. A56597), and if needed, adjust the focus or zoom to get bands as sharp as possible. Better quality pictures are obtained when the membrane is wet.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.